Substrate for western blotting, 1-Step™ NBT/BCIP
Catalog # PIER34042
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Conf:250 ml
- Descrizione:1-Step™ NBT/BCIP, ready to use
Specifications
About this item
The 1-Step NBT/BCIP is a one-component sensitive precipitating alkaline phosphatase substrate for immunoblotting, immunohistochemistry, and in situ hybridisation applications.