Orders LinkContactUsLinkComponent
Micrococcal nuclease solution (≥100 U/µl)
Micrococcal nuclease solution (≥100 U/µl)
Micrococcal Nuclease Solution (≥100 U/µL), 150 µl, 1
Codice catalogo PIER88216
Nome del fornitore:   Thermo Fisher Scientific
Micrococcal nuclease solution (≥100 U/µl)
Codice catalogo PIER88216
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  
Micrococcal nuclease solution (≥100 U/µl)
Micrococcal nuclease solution (≥100 U/µl)
Micrococcal Nuclease Solution (≥100 U/µL), 150 µl, 1
Numero CAS:  
Price
SFr. 296.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
This Micrococcal Nuclease (MNase) is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.
  • Enzyme digests nucleic acids (DNA and RNA)
  • Effective for double-stranded, single-stranded, circular and linear nucleic acids
  • Active in neutral to alkaline buffer conditions containing divalent calcium ions
  • Highest activity is toward single-stranded nucleic acid substrates with preference for AT or AU rich regions
Specifiche
Specifiche
  • Conf:
    1
  • Descrizione:
    Micrococcal Nuclease Solution (≥100 U/µL), 150 µl