Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Catalog # PIER15151
Supplier: Thermo Fisher Scientific
Specifications
- Confezione:5 plates per pack
- Colore:trasparente
- Descrizione:96-well strip microplate, 1×8 per strip
- Conf:5
Specifications
About this item
Biotin coated plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilisation of probes that are conjugated with streptavidin, avidin, or other biotin-binding proteins.
- Biotin group accessible for binding avidin or streptavidin
- Pre-blocked to reduce non specific binding
- Strip well plate format for the ultimate in convenience
- Coat volume: 200 µl
- Blocking volume: 300 µl