Hygiene sponge samplers
Collection sponges in poly envelopes, blu
TSCLTS/15-B
Sampling sponge dosed with neutralising buffer for the recovery of organisms including Listeria spp., Staphylococcus, Enterobacteriaceae such as salmonella.
Designed for the hygiene monitoring where chemical residue neutralisation is desired.
Packaging: Individual easy open stomacher pouch.
- Colore:blu
- Descrizione:Collection sponges in poly envelopes
- Seq:04