VWR®, Prepared Slides for Zoology: Vertebrates and Mammals
Catalog # 720-2928
Supplier: VWR Collection
Specifications
- Descrizione:Prepared slides for zoology
- Conf:1 KIT
Specifications
About this item
Prepared slides for zoology i.e. vertebrates and mammals including.
- Frog skin (c.s.), small intestine (c.s.), blood smear, testis (s.), liver (s.) and heart (s.)
- Rabbit liver (s.), testis (s.), vertebral column (c.s.), motor nerve endings (w.b.) and ovary (s.)
- Compact bone tissue
- Stomach walls (s.)
- Mammal small intestine (c.s.), large intestine (c.s.), pancreas, spleen (s.), gall bladder (s.), lung (s.), artery and vein (c.s.), kidney (l.s.) and eyeball (l.s.)
- Human sperm smear, skin (s.) through sweat gland and chromosome in blood, male and female
(w.b.) whole body; (s.) section; (l.s.) longitudinal section; (c.s.) cross section.