Immunoprecipitation, magnetic IP/Co-IP kit, c-Myc-tag, Pierce™
Codice catalogo PIER88844
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Specifiche
- Conf:1 KIT
- Descrizione:c-Myc-Tag Magnetic IP/Co-IP Kit, sufficient for 40 IP reactions using 25 µl of anti-c-Myc magnetic beads
Specifiche
Informazioni su questo articolo
The Pierce™ c-Myc-Tag Magnetic IP/Co-IP Kit contains specific immunoaffinity magnetic beads and reagents to perform immunoprecipitation assays of c-Myc fusion proteins or co-IP experiments using c-Myc-tagged bait proteins.
- Covalently immobilised high-quality anti-c-Myc monoclonal antibody enables high yields of immunoprecipitation products
- Stable, pre-blocked beads and specific antibody minimise off-target binding
- Complete kit and easy to follow instructions provide optimised protocols to perform IP or Co-IP experiments in approximately 1 hour
- Magnetic beads are compatible with manual and automated magnetic separation workflows
Delivery: The kit contains c-Myc-tagged positive control, 500 µl; anti-c-Myc magnetic beads, 1 ml; Mag c-Myc IP/Co-IP buffer-1, 50 ml; Mag c-Myc IP/Co-IP buffer-2 (20X), 20 ml; non-reducing sample buffer (5X), 5 ml; and elution buffer (pH 2,0), 5 ml.