Pegylation reagent, MT(PEG)₄ methyl-PEG-thiol compound
Référence PIER26132
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Spécifications
- Conf:100 mg
- Descrizione:MT(PEG)4
- Seq:01
Spécifications
À propos de cet article
Methyl-PEG4-thiol is a methyl- and sulfhydryl-terminated compound that contains a 4-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.
- PEG groups are flexible, non-immunogenic and hydrophilic
- Terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
- Terminal methyl group coats quantum dot and other surfaces to reduce nonspecific binding in assays
The use of MT(PEG)4 with CT(PEG)12 in surface modification can form a hydrophilic "lawn" of methyl ether-terminated PEGs with periodic exposed carboxylic acid-containing PEGs. The exposed carboxylic acid groups can be coupled to affinity ligands using the carbodiimide coupling reaction with EDC and sulpho-NHS.