Graphite spin columns, Pierce™
Catalog # PIER88302
Supplier: Thermo Fisher Scientific
Specifications
- Descrizione:Graphite spin columns, 30 columns
- Conf:1 KIT
Specifications
About this item
Pierce™ Graphite spin columns improve phosphopeptide analysis by efficiently binding hydrophilic peptides and efficiently removing urea, salts and other contaminants before mass spectrometry analysis.
- Convenient spin format for parallel processing of multiple samples
- High-binding capacity with recovery of up to 100 µg of hydrophilic peptides per column
- Porous graphite resin for cleaning up phosphopeptide samples before MS analysis
Enable fast and efficient capture, concentration, desalting and elution of hydrophilic peptides in less than ten minutes.