Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Catalog # PIER15031
Supplier: Thermo Fisher Scientific
Specifications
- Colore:trasparente
- Descrizione:96-well strip microplate assembled in frame, 1×8 per strip, flat bottom, corner notch
- Conf:100
- Volume pozzetti:360 µl
Specifications
About this item
PS, high antibody-binding 96-well microplate with individual 8-well strips in the plate frame for immunoassay applications.
- Clear polystyrene for colorimetric detections
- Plates are certified for performance, having consistently low coefficients of variation among wells
- Hydrophobic-ionic binding interaction
- Wells can be easily separated from strips with gentle twist
- Corner notch for use in standard plate readers