ContactUsLinkComponent
Homogeniser, FastPrep®-24, MP Biomedicals
Homogeniser, FastPrep®-24, MP Biomedicals
Catalog # ICNA116004500
Supplier:  MP Biomedicals
undefined
Homogeniser, FastPrep®-24, MP Biomedicals
Catalog # ICNA116004500
Supplier:  MP Biomedicals
Specifications
  • Descrizione:
    FastPrep®-24 benchtop homogeniser, 90-250V AC, 50/60 Hz, 1200 W
  • Conf:
    1
Specifications
About this item
FastPrep®-24 is used to homogenise biological samples. It lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
The FastPrep®-24 instrument is a high speed benchtop homogeniser offering the ultimate in speed and performance for the lysis of biological samples. Homogenisation of up to 24 samples takes place within 40 seconds. Uses a unique, optimised motion to disrupt cells through the multidirectional, simultaneous beating of specialised lysing matrix beads on the sample material. Developed for difficult and resistant samples.
Use with MP Biomedicals lysing matrix tubes or FastPrep® purification kits to lyse, grind or homogenise samples for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules.