Orders LinkContactUsLinkComponent
Stopcocks with GL thread connection, BOLA
Stopcocks with GL thread connection, BOLA
Supplier:   Bohlender
Stopcocks with GL thread connection, BOLA
Supplier:  Bohlender
Certificates
Stopcocks with GL thread connection, BOLA
Stopcocks with GL thread connection, BOLA
2 Options Available Below
Product Details & Documents
PTFE.
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −30 to +150 °C
  • Pressure resistant up to a maximum of 6 bar, vacuum suitable
Two-way stopcock with straight bore and one connection with GL thread or three-way stopcock with L-shaped bore and two connections with GL thread. Ideal for evacuating ground joint flasks or distributing liquid or gases with quick and easy disconnection of flow. Cylindrical stopcock plug for good leakage proof properties, stop valve with flow direction marking. Connection of tubing or tubes using laboratory screw joints.
Product Family Options
2-way stopcocks, straight
Product Information
  • Ø foro
    Filettatura
    Availability
    Price
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 162.00
    SFr. 162.00
    1 * 1 ST
       SFr. 162.00
3-way stopcocks, L-shaped bore
Product Information
  • Ø foro
    Filettatura
    Availability
    Price
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 256.00
    SFr. 256.00
    1 * 1 ST
       SFr. 256.00