Orders LinkContactUsLinkComponent
Pliers, Aesculap®
Supplier:   Aesculap
Pliers, Aesculap®
Supplier:  Aesculap
Certificates
Pliers, Aesculap®
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Descrizione
    Availability
    Price
  • Pliers, flat nosed, with serrated jaws, 170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Pliers, flat nosed, with cross grooves, 170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Pliers, flat nosed, 190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Pliers, round, with tapered jaws, 180 mm