Diethyl sebacate
Steel pail, 35 kg
Catalog # SPCMD2843-35KGBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate
- CAS Number:110-40-7
- Formula:C₁₄H₂₆O₄
- MDL Number:MFCD00009219
- Molecular Weight:258.36 g/mol
- Boiling Pt:312 °C (1013 hPa)
- Melting Pt:–3 °C
- Density:0,9646 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:203-764-5
Specifications
Specification Test Results
- Assay:98% Min.
Specifications
- Tipo di conf.:Steel pail
- Conf:35 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



