LDS-sample buffer (4X), non-reducing
Informazioni su questo articolo
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
2 varianti
Product Details & Documents
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- J61894.AC
- J61894.AD
- Aspetto:
- Blue
- Blue
- Forma fisica:
- Liquid
- Liquid