Orders LinkContactUsLinkComponent
Metals quality control standard for drinking water
Metals quality control standard for drinking water
Nome del fornitore:   Hach
Metals quality control standard for drinking water
Nome del fornitore:  Hach
Certificates
Metals quality control standard for drinking water
Metals quality control standard for drinking water
2 varianti
Product Details & Documents
These mixed standards are ready to use with no dilutions required. It include standards for determination of copper, iron and manganese.
Documents
Product Family Options
Informazioni sul prodotto
  • Concentrazione
    Conf
    Disponibilità
    Price
  • HACH2833649
    Metals quality control standard, high range
    Metals quality control standard, high range
    2,5 mg/l as Cu
    1,5 mg mg/l as Fe
    5,0 mg/l as Mn
    500 mL
    Log in per disponibilità
    Log in per disponibilità
    1 * 500 mL
    1 * 500 mL - SFr. 114.00
    SFr. 114.00
    1 * 500 mL
       SFr. 114.00
  • HACH2833749
    Metals quality control standard, low range
    Metals quality control standard, low range
    1.0 mg/l as Cu
    0,3 mg mg/l as Fe
    0,1 mg/l as Mn
    500 mL
    Log in per disponibilità
    Log in per disponibilità
    1 * 500 mL
    1 * 500 mL - SFr. 114.00
    SFr. 114.00
    1 * 500 mL
       SFr. 114.00