Orders LinkContactUsLinkComponent
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Nome del fornitore:   WTW
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Nome del fornitore:  WTW
Certificates
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
5 varianti
Product Details & Documents
This compact portable multi-parameter instrument enables measurements of pH, conductivity and dissolved oxygen with intelligent digital sensors (IDS). It has a universal measurement channel for different parameters. The robust, waterproof design makes it ideal for measurements in the field and in the laboratory.
  • Well laid out menus make operation safe and easy
  • MultiLab® Importer software enables data acquisition via Excel®
  • QSC function for optimum tracking of sensor quality
  • Calibration records and additional information are stored in the sensor
  • Wireless-ready for IDS wireless modules (for cable-free operation)
  • Available as sets in cases with sensors and accessories
Delivery information: Single meter (without electrodes) is supplied with case, MultiLab® Importer software, batteries, USB cable and short manual. The sets (except for set U) additionally include sensors.
Set U includes: Meter, case, instruction manual, stand, beaker, buffer 4 and 7, 3 mol/l KCl, conductivitiy standard, CD-ROM, driver software for USB, rechargeable batteries, USB cable, power supply, armoring SM Pro.
Please note: A wireless module is required for use with the wireless electrodes. This module is not included and needs to be ordered separately.
Documents
Specifications for Product Family
Specifiche
  • Catalog No:
  • 662-2135
  • 662-2136
  • 662-2137
  • 662-2138
  • 662-2139
  • Modello:
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Display:
  • LCD grafico, retroilluminato
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Alimentazione:
  • 4 AA da 1,5 V (incluse); 4 NiMH-Akku da 1,2 V o tramite porta USB
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • Conformità:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Capacità della memoria:
  • 500 (manuale); 4500 (automatico)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • Interfacce:
  • 1 ingresso per misurazione
  • 1 measurement input
  • 1 measurement input
  • 1 measurement input
  • 1 measurement input
  • Uscite:
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
Specifiche
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • 662-2135
    Misuratore multiparametro, Multi 3510 IDS, solo misuratore (senza elettrodo)
    Misuratore multiparametro, Multi 3510 IDS, solo misuratore (senza elettrodo)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 1’490.00
    SFr. 1’490.00
    1 * 1 ST
       SFr. 1’490.00
    Specifiche:
    More
    Specifications
    Cat. No.
    662-2135
    Modello
    Multi 3510 IDS
    Display
    LCD grafico, retroilluminato
    Alimentazione
    4 AA da 1,5 V (incluse); 4 NiMH-Akku da 1,2 V o tramite porta USB
    Conformità
    IP 67
    Capacità della memoria
    500 (manuale); 4500 (automatico)
    Interfacce
    1 ingresso per misurazione
    Uscite
    Mini USB-B
  • 662-2136
    Multi-parameter meter, handheld, Multi 3510 IDS, set 1, with SenTix® 940 (pH gel electrode)
    Multi-parameter meter, handheld, Multi 3510 IDS, set 1, with SenTix® 940 (pH gel electrode)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 1’910.00
    SFr. 1’910.00
    1 * 1 ST
       SFr. 1’910.00
    Specifiche:
    More
    Specifications
    Cat. No.
    662-2136
    Modello
    Multi 3510 IDS
    Display
    Graphic LCD, backlit
    Alimentazione
    4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
    Conformità
    IP 67
    Capacità della memoria
    500 (manual); 4500 (automatic)
    Interfacce
    1 measurement input
    Uscite
    Mini USB-B
  • 662-2137
    Multi-parameter meter, handheld, Multi 3510 IDS, set 3, with TetraCon® 925 (4-electrode conductivity cell)
    Multi-parameter meter, handheld, Multi 3510 IDS, set 3, with TetraCon® 925 (4-electrode conductivity cell)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 2’370.00
    SFr. 2’370.00
    1 * 1 ST
       SFr. 2’370.00
    Specifiche:
    More
    Specifications
    Cat. No.
    662-2137
    Modello
    Multi 3510 IDS
    Display
    Graphic LCD, backlit
    Alimentazione
    4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
    Conformità
    IP 67
    Capacità della memoria
    500 (manual); 4500 (automatic)
    Interfacce
    1 measurement input
    Uscite
    Mini USB-B
  • 662-2138
    Multi-parameter meter, handheld, Multi 3510 IDS, set 4, with FDO® 925 (optical DO sensor)
    Multi-parameter meter, handheld, Multi 3510 IDS, set 4, with FDO® 925 (optical DO sensor)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 2’900.00
    SFr. 2’900.00
    1 * 1 ST
       SFr. 2’900.00
    Specifiche:
    More
    Specifications
    Cat. No.
    662-2138
    Modello
    Multi 3510 IDS
    Display
    Graphic LCD, backlit
    Alimentazione
    4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
    Conformità
    IP 67
    Capacità della memoria
    500 (manual); 4500 (automatic)
    Interfacce
    1 measurement input
    Uscite
    Mini USB-B
  • 662-2139
    Multi-parameter meter, handheld, Multi 3510 IDS, set U (without electrode), with case, accessories and armor SM Pro
    Multi-parameter meter, handheld, Multi 3510 IDS, set U (without electrode), with case, accessories and armor SM Pro
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 1’760.00
    SFr. 1’760.00
    1 * 1 ST
       SFr. 1’760.00
    Specifiche:
    More
    Specifications
    Cat. No.
    662-2139
    Modello
    Multi 3510 IDS
    Display
    Graphic LCD, backlit
    Alimentazione
    4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
    Conformità
    IP 67
    Capacità della memoria
    500 (manual); 4500 (automatic)
    Interfacce
    1 measurement input
    Uscite
    Mini USB-B
Electrodes
Informazioni sul prodotto
  • Codice catalogo
    Concentrazione
    Descrizione
    Tipo
    Connettore
    Ø×L
    Materiale
    Temperatura
    Disponibilità
    Price
  • 0,00 - 20,00 mg/L
    Sonda ossigeno disciolto IDS FDO® 925
    FDO® 925
    Cavo da 1,5 m
    15,3×140 mm
    POM, acciaio inox
    0...+50,0 °C
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 1’600.00
    SFr. 1’600.00
    1 * 1 ST
       SFr. 1’600.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Foro
    Polimero
    0...+60 °C
    Elettrodo per pH IDS, resistente alla pressione (10 bar), SensoLyt® 900-P, predisposizione wireless
    12×120 mm
    Vetro
    SensoLyt® 900-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 820.00
    SFr. 820.00
    1 * 1 ST
       SFr. 820.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Ceramica
    KCl 3 mol/l
    0...+80 °C
    Elettrodo per pH IDS, SenTix® 950-P, predisposto per wireless
    12×120 mm
    Plastica
    SenTix® 950-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 508.00
    SFr. 508.00
    1 * 1 ST
       SFr. 508.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Filo di platino
    KCl 3 mol/l
    0...+100 °C
    Elettrodo per pH IDS, SenTix® 980-P, predisposto per wireless
    12×120 mm
    Vetro
    SenTix® 980-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 509.00
    SFr. 509.00
    1 * 1 ST
       SFr. 509.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • 3 in ceramica
    Gel
    0...+80 °C
    Elettrodo per pH IDS, SenTix® 945-P, predisposto per wireless
    12×120 mm
    Vetro
    SenTix® 945-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 541.00
    SFr. 541.00
    1 * 1 ST
       SFr. 541.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Anello apribile
    KCl 3 mol/l
    0...+60 °C
    Elettrodo per pH IDS, SenTix® HW-T-900-P, predisposto per wireless
    12×120 mm
    Vetro
    SenTix® HW-T-900-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 559.00
    SFr. 559.00
    1 * 1 ST
       SFr. 559.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Foro
    Polimero
    0...+50 °C
    Elettrodo per pH IDS, inserzione, SenTix® Sp-T900-P, predisposizione wireless
    15/5×65/25 mm
    Plastica
    SenTix® Sp-T 900-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 652.00
    SFr. 652.00
    1 * 1 ST
       SFr. 652.00
Informazioni sul prodotto
  • Codice catalogo
    Concentrazione
    Temperatura
    Tipo
    Ø×L
    Descrizione
    Connettore
    Materiale
    Disponibilità
    Price
  • Foro
    Polimero
    Vetro
    12×120 mm
    Elettrodo IDS ORP, resistente alla pressione (10 bar), SensoLyt® ORP 900-P, predisposizione wireless
    Testa a spina per cavo AS IDS o modulo wireless
    0...+60 °C
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 975.00
    SFr. 975.00
    1 * 1 ST
       SFr. 975.00
Informazioni sul prodotto
  • Codice catalogo
    Concentrazione
    Temperatura
    Tipo
    Ø×L
    Descrizione
    Connettore
    Materiale
    Disponibilità
    Price
  • Ceramica
    3 mol/l KCl
    Vetro
    12×120 mm
    Elettrodo IDS ORP, SenTix® ORP-T 900-P, predisposizione wireless
    Testa a spina per cavo AS IDS o modulo wireless
    0...+60 °C
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 468.00
    SFr. 468.00
    1 * 1 ST
       SFr. 468.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Fibra
    Gel
    0...+80 °C
    Elettrodo per pH IDS, SenTix® 940-P, predisposto per wireless
    12×120 mm
    Plastica
    SenTix® 940-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 419.00
    SFr. 419.00
    1 * 1 ST
       SFr. 419.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Filo di platino
    KCl 3 mol/l
    0...+100 °C
    Elettrodo per pH IDS, SenTix® Micro 900-P, predisposto per wireless
    12/5×70/130 mm
    Vetro
    SenTix® Micro 900-P
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 894.00
    SFr. 894.00
    1 * 1 ST
       SFr. 894.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Fibre
    Gel
    0...+80 °C
    Elettrodo IDS pH Sentix® 940
    12×120 mm
    Plastica
    Sentix® 940
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 419.00
    SFr. 419.00
    1 * 1 ST
       SFr. 419.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Fibre
    Gel
    0...+80 °C
    Elettrodo IDS pH Sentix® 940-3
    12×120 mm
    Plastica
    Sentix® 940-3
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 474.00
    SFr. 474.00
    1 * 1 ST
       SFr. 474.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Ceramica
    3M KCl
    0...+80 °C
    Elettrodo IDS pH Sentix® 950
    12×120 mm
    Plastica
    Sentix® 950
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 508.00
    SFr. 508.00
    1 * 1 ST
       SFr. 508.00
Informazioni sul prodotto
  • Codice catalogo
    Ø×L
    Descrizione
    Temperatura
    Concentrazione
    Connettore
    Materiale
    Tipo
    Disponibilità
    Price
  • Filo di platino
    3M KCl
    0...+100 °C
    Elettrodo IDS pH Sentix® 980
    12×120 mm
    Vetro
    Sentix® 980
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 474.00
    SFr. 474.00
    1 * 1 ST
       SFr. 474.00
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Disponibilità
    Price
  • AS/IDS-1,5, cavo di collegamento per sensori IDS con testa a spina impermeabile, lunghezza 1,5 m
    AS/IDS-1,5, cavo di collegamento per sensori IDS con testa a spina impermeabile, lunghezza 1,5 m
    662-1733
    1,5 m
    Log in per disponibilità
    1 * 1 ST - SFr. 163.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Cavo adattatore, 1,5 m, per il collegamento di un elettrodo SenTix® combinato con testa a spina S7 a uno strumento MultiLine®
    Cavo adattatore, 1,5 m, per il collegamento di un elettrodo SenTix® combinato con testa a spina S7 a uno strumento MultiLine®
    662-1863
    Log in per disponibilità
    1 * 1 ST - SFr. 411.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Conduttanza
    Disponibilità
    Price
  • IDS conductivity cell, TetraCon® 925, 4-cell graphite
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
    663-0152
    TetraCon® 925, 4 elettrodi, grafite
    10 µS/cm - 2000 mS/cm
    Log in per disponibilità
    1 * 1 ST - SFr. 911.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Kit QSC, kit di calibrazione iniziale per elettrodi pH IDS (scatola con 3 fiale: pH 4,01/pH 6,86/pH 9,18)
    Kit QSC, kit di calibrazione iniziale per elettrodi pH IDS (scatola con 3 fiale: pH 4,01/pH 6,86/pH 9,18)
    662-2132
    Log in per disponibilità
    1 * 1 ST - SFr. 55.10
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Grande scatola di campo, KS MultiLine® 2, senza misuratore/sensori, incl. rivestimento SM Pro, buffer STP 4 e STP 7, supporto, beaker, conducibilità standard 1413 µS/cm a 25 °C
    Grande scatola di campo, KS MultiLine® 2, senza misuratore/sensori, incl. rivestimento SM Pro, buffer STP 4 e STP 7, supporto, beaker, conducibilità standard 1413 µS/cm a 25 °C
    662-2134
    Log in per disponibilità
    1 * 1 ST - SFr. 468.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Scatola, KS MultiLine® 1, senza misuratore/sensori, incl. rivestimento SM Pro, buffer STP 4 e STP 7, supporto, beaker, conducibilità standard 1413 µS/cm a 25 °C
    Scatola, KS MultiLine® 1, senza misuratore/sensori, incl. rivestimento SM Pro, buffer STP 4 e STP 7, supporto, beaker, conducibilità standard 1413 µS/cm a 25 °C
    662-2133
    Log in per disponibilità
    1 * 1 ST - SFr. 417.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Custodia protettiva SM PRO per ProfiLine / MultiLine
    Custodia protettiva SM PRO per ProfiLine / MultiLine
    662-1278
    Log in per disponibilità
    1 * 1 ST - SFr. 142.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Modulo wireless/IDS per sensori con testa a spina, IDS WLM-S
    Modulo wireless/IDS per sensori con testa a spina, IDS WLM-S
    665-0423
    Log in per disponibilità
    1 * 1 ST - SFr. 291.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Modulo wireless/IDS per misuratori IDS, IDS WLM-M
    Modulo wireless/IDS per misuratori IDS, IDS WLM-M
    665-0424
    Log in per disponibilità
    1 * 1 ST - SFr. 234.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Caricatore USB per moduli wireless
    Caricatore USB per moduli wireless
    665-0425
    Log in per disponibilità
    1 * 1 ST - SFr. 197.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Kit composto da modulo wireless per sensore e metro (1 pz), caricatore USB e alimentatore USB universale
    Kit composto da modulo wireless per sensore e metro (1 pz), caricatore USB e alimentatore USB universale
    662-2050
    Log in per disponibilità
    1 * 1 ST - SFr. 639.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Alimentatore USB universale
    Alimentatore USB universale
    662-2148
    Log in per disponibilità
    1 * 1 ST - SFr. 55.30