Industrial gloves, uvex phynomic C3
Supplier: UVEX
Certificates
About this item
The uvex phynomic is an ultra-light all-round safety glove with good cut protection (Cut 3). The polyamide/elastane/HPPE/glass glove liners feature an aqua-polymer foam coating on palm and fingertips.
- Foam coating is extremely flexible, provides good grip and leaves no trace on sensitive surfaces
- Excellent dexterity
- Very high level of breathability
- Accelerator-free and suitable for allergy sufferers
Ideal for precision assembly and precision work. Suitable for use in dry and slightly damp areas.
Certifications: EN 388:2016 (4X43B)
Food contact approved
OEKO-Tex® Standard 100
Food contact approved
OEKO-Tex® Standard 100
Specifications
- Catalog No:
- 112-4947
- 112-4945
- 112-4946
- 112-4944
- 112-4943
- 112-4942
- Modello:
- C3
- C3
- C3
- C3
- C3
- C3
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Finitura:
- Aqua-polymer foam coating on palm and fingertips
- Aqua-polymer foam coating on palm and fingertips
- Aqua-polymer foam coating on palm and fingertips
- Aqua-polymer foam coating on palm and fingertips
- Aqua-polymer foam coating on palm and fingertips
- Aqua-polymer foam coating on palm and fingertips
- Tipo polsino:
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Norma EN:
- EN 388:2016 (4X43B)
- EN 388:2016 (4X43B)
- EN 388:2016 (4X43B)
- EN 388:2016 (4X43B)
- EN 388:2016 (4X43B)
- EN 388:2016 (4X43B)
- Approvati per il contatto con gli alimenti:
- +
- +
- +
- +
- +
- +