Anti-LMAN2L Rabbit Polyclonal Antibody
Catalog # ABGEAP20404B
Supplier: Abgent
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:lectin, mannose-binding 2-like
- Simbolo dell'antigene:LMAN2L
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Epitopo:C-terminal
- ID gene:81562
- Sinomini antigene:VIPL
- Numero amminoacido:322 to 348
- Tampone di conservazione:pH 7,4
- Peso molecolare:39.711 kDa
- Sequenza:CNKWQEQSRKRFY
- Temperatura di conservazione:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentrazione:0.63
- Temperatura di spedizione:4 °C
- Immunogeno:This LMAN2L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 322-348 amino acids from the C-terminal region of human LMAN2L.
- Purificazione:Protein A & Peptide affinity purification
- Conf:400 µl
Specifications
About this item
Type: Primary
Antigen: LMAN2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human