Anti-MS4A8 Rabbit Polyclonal Antibody
Catalog # ABGEAP18101A
Supplier: Abgent
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Membrane-spanning 4-Domains, Subfamily A, Member 8
- Simbolo dell'antigene:MS4A8
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Epitopo:N-terminal
- ID gene:83661
- Sinomini antigene:CD20L5|MS4A4|4SPAN4|MS4A8B
- Numero amminoacido:44 to 70
- Tampone di conservazione:pH 7,4
- Peso molecolare:26.29 kDa
- Sequenza:GQPVQKALKEGKC
- Temperatura di conservazione:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentrazione:0.67
- Temperatura di spedizione:4 °C
- Immunogeno:This MS4A8B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 44-70 amino acids from the N-terminal region of human MS4A8B.
- Purificazione:Protein A & Peptide affinity purification
- Conf:400 µl
Specifications
About this item
Type: Primary
Antigen: MS4A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human