Serological pipettes, Shortie, Nunc™
Catalog # 612-4741
Supplier: Thermo Fisher Scientific
Specifications
- Confezione:Individually wrapped; 50/bag; 200/case
- Sterili:+
- Capacità:10 ml
- Seq:02
- Conf:200
Specifications
About this item
PS, transparent, graduated and sterile. These sleek pipettes, made of ultra-clear virgin PS, are ideal for easy manoeuvring under the hood.
- Disposable, plugged
- Certified non pyrogenic
- Bold clear graduations with easy to read scale markings
- Non drip tip for pipette precision
- Sterility assurance level SAL 10⁻⁶
Packaging: Individually wrapped in paper/plastic peel packaging.