Reagent reservoirs
Codice catalogo 613-3093
Nome del fornitore: Sorenson Bioscience
Specifiche
- Confezione:Individually wrapped
- Sterili:+
- Tipo:V-bottom
- Capacità:25 ml
- Colore:bianco
- Seq:01
- Conf:100
Specifiche
Informazioni su questo articolo
These reagent reservoirs made of PS or PVC are designed to accommodate all types of pipettors from single channel to 16-channel.
- Made of FDA grade plastic
- Graduation marks on interior wall
- Lot certified Rnase/Dnase-free and non pyrogenic
- Pour-off spouts in each corner
- Availabe non sterile or sterile; sterility validated at 10⁻⁶