Safety shoes, lace-up, Step One, Sensation
Codice catalogo UPOWSO20165/43
Nome del fornitore: U-Power
Specifiche
- Taglia scarpe:43
- Colore:blu/nero
- Conf:1 PAIR
Specifiche
Informazioni su questo articolo
Safety shoes made from soft suede leather with textile inserts and a slip-resistant dual-density PU sole.
- Comfortable, anatomically shaped ergo dry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- Air toe protective composite toecap with breathable membrane (200 J)
- Puncture-resistant steel midsole
Colour: Blue
Certifications: EN 20345:2011; S1P, SRC