Adapters, sperical joints
For reaction vessels, 316 stainless steel, thread length 14 mm.
For reaction vessels, 316 stainless steel, thread length 14 mm.
Product Family Options
- Tipo
- Socket S 35 with steel thread M 33×1,5, O.Ø 30×2,4 mm, I.Ø 25,2 mm1 * 1 STSFr. 835.00
- Socket S 51 with steel thread M 48×1,5, O.Ø 36×2,4 mm, I.Ø 31,2 mm1 * 1 STSFr. 1’040.00