Spatula
300 mm, 188 mm, 50 ml
Codice catalogo 129-1158
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
This spatula is made of premium stainless steel, high-gloss polished and completely welded, thus completely free of undercuts.
- Very sturdy
- Easy to clean
- Autoclavable, can be flame-sterilised
- Can pierce solid substances or sacks and BigBags
Ideal for taking samples of powders, granulates and pasty media, especially in food and pharmaceutical applications.
Specifiche
- Capacità:50 ml
- Lunghezza:300 mm
- Lunghezza lama:188 mm