Anti-FLAG-tag Mouse Monoclonal Antibody [clone: FG4R]
AGRIAS152871
: Agrisera
- Tipo anticorpo:Primary
- Tipo di anticorpi:FLAG tag
- Simbolo dell'antigene:FLAG
- Clonalità:Monoclonale
- Clone:FG4R
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Reattività:Ficus glabrata
- Western blot:Yes
- Immunogeno:KLH-conjugated DYKDDDDK (FLAG) synthetic peptide
- Applicazioni testate:Dot Blot
- Taglia:50 µg
- Conf:50 µG
FLAG-tag is a tag that can be added to a protein sequence motif DYKXXD. It can be used for affinity chromatography, to separate recombinant protein from wt proteins. It can also be used to isolate protein complexes with multiple subunits.,FLAG-tag is a tag that can be added to a protein sequence motif DYKXXD. It can be used for affinity chromatography, to separate recombinant protein from wt proteins. It can also be used to isolate protein complexes with multiple subunits.
Type: Primary
Antigen: FLAG
Clonality: Monoclonal
Clone: FG4R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Ficus glabrata