Order Entry
Switzerland
ContactUsLinkComponent
Anti-GIT1 Rabbit Polyclonal Antibody
Anti-GIT1 Rabbit Polyclonal Antibody
Codice catalogo BOSSBS-5372R
Nome del fornitore:  Bioss
Anti-GIT1 Rabbit Polyclonal Antibody
Codice catalogo BOSSBS-5372R
Nome del fornitore:  Bioss

Specifiche

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    G Protein-Coupled Receptor Kinase Interacting ArfGAP 1
  • Simbolo dell'antigene:
    GIT1
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Forma:
    liquid
  • ID gene:
    28964
  • Sinomini antigene:
    ARF GTPase-activating protein GIT1|GIT16|Cat-1|G protein-coupled receptor kinase-interactor 1|GRK-interacting protein 1|Cool-associated and tyrosine-phosphorylated protein 1
  • Modifica:
    Ser375
  • Tampone di conservazione:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentrazione:
    1 ug/ul
  • Temperatura di spedizione:
    4 °C
  • Purificazione:
    Purified by Protein A.
  • Taglia:
    100 μg
  • Conf:
    100 µl

Specifiche

Informazioni su questo articolo

Heterotrimeric G protein-mediated signal transduction is a dynamically regulated process with the intensity of signal decreasing over time despite the continued presence of the agonist (1,2). G protein-coupled receptor kinases (GRKs) are activated by activated G protein-coupled receptors, and they function to phosphorylate and inactivate cell surface receptors in the heterotrimeric G protein signaling cascade (3,4). GIT1 (for GRK-interactor 1) and GIT2 are GTPase-activating proteins (GAP) for members of the ADP ribosylation factor (ARF) family of small GTP-binding proteins, which are involved in vesicular trafficking (5,6). GIT1 overexpression results in reduced internalization and resensitization of b2-adrenergic receptor, thus reducing b2-adrenergic receptor signaling (5).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human