Order Entry
Switzerland
ContactUsLinkComponent
Anti-GIT1 Rabbit Polyclonal Antibody
Anti-GIT1 Rabbit Polyclonal Antibody
N° de catalogue BOSSBS-4110R
Fournisseur:  Bioss
undefined
Anti-GIT1 Rabbit Polyclonal Antibody
N° de catalogue BOSSBS-4110R
Fournisseur:  Bioss

Spécifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    G Protein-Coupled Receptor Kinase Interacting ArfGAP 1
  • Simbolo dell'antigene:
    GIT1
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Forma:
    liquid
  • ID gene:
    28964
  • Sinomini antigene:
    GIT1|ARF GTPase-activating protein GIT1|CAT1|G protein-coupled receptor kinase-interactor 1|GRK-interacting protein 1|CAT-1|ARF GAP GIT1|Cool-associated and tyrosine-phosphorylated protein 1
  • Tampone di conservazione:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentrazione:
    1 ug/ul
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    480-530/761
  • Purificazione:
    Purified by Protein A.
  • Taglia:
    100 μg
  • Conf:
    100 µl

Spécifications

À propos de cet article

GTPase-activating protein for the ADP ribosylation factor family. May serve as a scaffold to bring together molecules to form signaling modules controlling vesicle trafficking, adhesion and cytoskeletal organization. Increases the speed of cell migration, as well as the size and rate of formation of protrusions, possibly by targeting PAK1 to adhesions and the leading edge of lamellipodia. Sequesters inactive non-tyrosine-phosphorylated paxillin in cytoplasmic complexes. Involved in the regulation of cytokinesis; the function may involve SDCCAG3 and PTPN13 (By similarity).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat