Orders LinkContactUsLinkComponent
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®
Certificates
pH buffer solutions, Specpure®
19 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Product Family Options
Product Information
  • Valore pH
    Accuratezza pH
    Tipo di tampone pH
    Colore
    Temperatura di riferimento
    Tipo di conf.
    Conf
    Availability
    Price
  • 1,00
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 57.50 SFr.
    57.50 SFr.
    1 * 500 mL
       57.50 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    40446.AP
  • 1,00
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 89.70 SFr.
    89.70 SFr.
    1 * 1 L
       89.70 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    40446.K2
  • 1,00
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 159.00 SFr.
    159.00 SFr.
    1 * 4 L
       159.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    40446.K7
  • 1,68
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59.30 SFr.
    59.30 SFr.
    1 * 500 mL
       59.30 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    42415.AP
  • 1,68
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 93.20 SFr.
    93.20 SFr.
    1 * 1 L
       93.20 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    42415.K2
  • 1,68
    ±0,01
    HCl/KCl
    incolore
    25 °C
    bottiglia in vetro
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 165.00 SFr.
    165.00 SFr.
    1 * 4 L
       165.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    42415.K7
  • 4,00
    ±0,01
    ftalato
    rosso
    25 °C
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59.30 SFr.
    59.30 SFr.
    1 * 500 mL
       59.30 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38713.AP
  • 4,00
    ±0,01
    ftalato
    rosso
    25 °C
    Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 93.00 SFr.
    93.00 SFr.
    1 * 1 L
       93.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38713.K2
  • 4,00
    ±0,01
    ftalato
    rosso
    25 °C
    Glass bottle
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 164.00 SFr.
    164.00 SFr.
    1 * 4 L
       164.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38713.K7
  • 4,00
    ±0,01
    ftalato
    rosso
    25 °C
    Plastic container
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L - 634.00 SFr.
    634.00 SFr.
    1 * 20 L
       634.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38713.M4
  • 7,00
    ±0,01
    fosfato
    verde
    25 °C
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59.90 SFr.
    59.90 SFr.
    1 * 500 mL
       59.90 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38711.AP
  • 7,00
    ±0,01
    fosfato
    verde
    25 °C
    Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 94.20 SFr.
    94.20 SFr.
    1 * 1 L
       94.20 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38711.K2
  • 7,00
    ±0,01
    fosfato
    verde
    25 °C
    Glass bottle
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 166.00 SFr.
    166.00 SFr.
    1 * 4 L
       166.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38711.K7
  • 10,00
    ±0,01
    Carbonate
    incolore
    25 °C
    bottiglia in vetro
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 60.40 SFr.
    60.40 SFr.
    1 * 500 mL
       60.40 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38710.AP
  • 10,00
    ±0,01
    Carbonate
    incolore
    25 °C
    bottiglia in vetro
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 94.90 SFr.
    94.90 SFr.
    1 * 1 L
       94.90 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38710.K2
  • 10,00
    ±0,01
    Carbonate
    incolore
    25 °C
    bottiglia in vetro
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 175.00 SFr.
    175.00 SFr.
    1 * 4 L
       175.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38710.K7
  • 10,00
    ±0,01
    Carbonate
    blu
    25 °C
    bottiglia in vetro
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 58.40 SFr.
    58.40 SFr.
    1 * 500 mL
       58.40 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38709.AP
  • 10,00
    ±0,01
    Carbonate
    blu
    25 °C
    bottiglia in plastica
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 91.70 SFr.
    91.70 SFr.
    1 * 1 L
       91.70 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38709.K2
  • 10,00
    ±0,01
    Carbonate
    blu
    25 °C
    bottiglia in vetro
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 162.00 SFr.
    162.00 SFr.
    1 * 4 L
       162.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    38709.K7