Functional larynx model
Codice catalogo BINH1013870
Nome del fornitore: 3B Scientific
Specifiche
- Descrizione:Functional larynx model
- Ingrandimento:2,5×
- Conf:1
Specifiche
Informazioni su questo articolo
The epiglottis, vocal cords and arytenoid cartilage are movable in the functional larynx. Additionally the following structures of the human larynx are represented:
- Hyoid bone
- Cricoid cartilage
- Thyroid cartilage
- Thyroid
- Parathyroid glands
This anatomically correct human larynx is a great addition to any doctors office or classroom.
Delivery: On a stand.