HPLC columns, Acclaim™ AmG C18
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
These columns are designed for ion pairing reversed phase HPLC (IP-RPLC) analysis of aminoglycoside antibioticss, including drug purity characterisation and quantification, therapeutic drug monitoring and residual control testing in meat / poultry. The Acclaim™ AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions.
- Great selectivity and high resolution for the aminoglycoside antibiotics analysis
- Stable at low pH, high temperature, organic solvents and aqueous mobile phases
- Can be used with UV, CAD, ELSD detectors
Specifiche
- Catalog No:
- 555-7432
- 555-7431
- 555-7430
- 555-7429
- 555-7433
- 555-7428
- Pressione max.:
- 8000 psi
- 8000 psi
- 8000 psi
- 8000 psi
- 8000 psi
- 8000 psi
- Range di pH:
- 0,5 - 10
- 0,5 - 10
- 0,5 - 10
- 0,5 - 10
- 0,5 - 10
- 0,5 - 10
- Limite di temperatura:
- <80 °C
- <80 °C
- <80 °C
- <80 °C
- <80 °C
- <80 °C