Heating tapes, TeMS2
Nome del fornitore: ISOPAD
Vai a
Informazioni su questo articolo
Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.
- Temperature resistant up to 260 °C
- Non hazardous and used in ordinary areas
- Parallel arrangement ensures a wide heat transfer surface
IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.
9 varianti
Product Details & Documents
Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.
- Temperature resistant up to 260 °C
- Non hazardous and used in ordinary areas
- Parallel arrangement ensures a wide heat transfer surface
IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.
Product Family Options
TeMS2
Informazioni sul prodotto
- DisponibilitàPrice
- ISOP584216-000Heating tape TeMS2, 21,1 m length, 50 W
- ISOP411376-000Heating tape TeMS2, 22,2 m length, 100 W
- ISOP067444-000Heating tape TeMS2, 23,5 m length, 140 W
TeMS4
Informazioni sul prodotto
- DisponibilitàPrice
- ISOP458786-000Heating tape TeMS4, 1,1 m length, 100 W
- ISOP124922-000Heating tape TeMS4, 5,50 m length, 490 W
- ISOP927204-000Heating tape TeMS4, 14 m length, 1260 W
TeMS6
Informazioni sul prodotto
- DisponibilitàPrice
- ISOP184778-000Heating tape TeMS6, 63 m length, 490 W
- 454-2192Heating tape TeMS6, 62 m length, 330 W
- 454-2191Heating tape TeMS6, 61 m length, 160 W