Folded towels, ZetForm®
Catalog # ZVGA15301-04
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Specifications
- Confezione:2400 towels/pack (20×120)
- Colore:naturale
- Descrizione:C-fold towels, 1 ply
- L×L:500×250 mm
- Seq:01
- Conf:2.400
Specifications
About this item
These 1 ply towels dry hands quickly and reliably. Ideal for use in washrooms with high frequencies: municipal facilities, restaurants, medical environments and more.
- 100% recycling material
- High absorbency, robust
- C-fold
Colour: Natural