Hand dynamometer, Smedley spring-type, Baseline®
Baseline® Smedley spring dynamometers, 0 - 90 kg
Catalog # BINHW54654
Marketsource
Supplier: 3B Scientific
Hand dynamometer, Smedley spring-type, Baseline®
Catalog # BINHW54654
Supplier: 3B Scientific
Marketsource
Jump to:
Product Details & Documents
Digital, lightweight Smedley spring dynamometer offers accurate grip strength readout.
- 5-preset grip sizes
- Captures and displays maximum grip force in both kilogrammes and pounds
- Unit assesses results according to age and gender
- Unit saves/stores results for fast retrieval with up to 19 users
- The adjustable handle comfortably accommodates any hand size.
: Supplied with protective carrying case.
Specifications
- Range di misura:0 - 90 kg
- Descrizione:Baseline® Smedley spring dynamometers