Lens cleaning paper
N° de catalogue NEOL1-7218
Fournisseur: NEOLAB
Spécifications
- Descrizione:Lens cleaning paper
- L×L:130×100 mm
- Conf:250
Spécifications
À propos de cet article
Cleaning paper suitable for use with lenses, slides, cuvettes, glass plates, and more.
- Cleans without scratching
- Dust-free and fibre-free