EGC monitoring electrode, Red Dot™
MMMEM2268-3
- Ø×L:Ø 32 mm
- Descrizione:Soft cloth monitoring electrode, with conductive adhesive and lift tab
- Seq:01
- Conf:600
These monitoring electrodes feature a soft cloth backing and gentle adhesives which makes them ideal for use on fragile skin and on the tender skin of younger pediatric patients.
- Gentle adhesive reduces risk of skin trauma
- Smaller footprint for children: Accommodates smaller chest sizes
- Soft cloth backing provides a low-profile adhesion which can minimise potential premature edge lift when used around medical devices or fabrics
- Conforms well to the skin enabling patient comfort