Dual plate rotators
Catalog # HARV74-2334
Supplier: HARVARD APPARATUS
Specifications
- Descrizione:Dual plate rotator
- Conf:1
Specifications
About this item
Dual plate rotator with variable rotation rates, can be used with the Harvard apparatus 96-well equilibrium dialyser. The rotator speeds up the equilibrium dialysis process by keeping the sample in constant motion thereby ensuring higher reproducibility of results.