ContactUsLinkComponent
Qualitative filter papers, grades MN 618 and 618 1/4, circles and folded filters
undefined
Qualitative filter papers, grades MN 618 and 618 1/4, circles and folded filters
  • Catalog No:
  • MANA436009
  • MANA436024
  • MANA536018
  • MANA436005
  • MANA536015
  • MANA436018
  • MANA536024
  • MANA436015
  • MANA536012
  • MANA436032
  • MANA536032
  • MANA436011
  • MANA436007
  • MANA436012
  • Peso:
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • Spessore:
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • Velocità di filtrazione:
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • 22 s (DIN 53137)
  • Contenuto di cenere:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
Frequently Bought Together