Gas detection tubes for ethylene oxide
Supplier: Dräger
Certificates
About this item
To measure the 'spot concentrations' of ethylene oxide.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Specifications
- Catalog No:
- 151-0108
- 118-0292
- Modello:
- Ethylene oxide 25/a
- Ethylene oxide 1/a
- Modifica del colore:
- Pale yellow to turquoise green
- White to pink
- Umidità:
- 3 - 15 mg/l H₂O
- 3 - 15 mg/l H₂O
- Tempo di misurazione:
- 6 minutes
- 8 minutes
- N. di corse:
- 30
- 20
- Deviazione standard:
- ±20 - 30 %
- ±20 - 30 %
- Gamma standard:
- 25 - 500 ppm
- 1 - 15 ppm
- Range di temperatura:
- 15…30 °C
- 10…30 °C