Orders LinkContactUsLinkComponent
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system
DirectPCR® DNA Lysis Reagents offer a quick and cost-effective 'single-tube' system for the rapid preparation of PCR template DNA from mouse tails, ears, embryonic material or mammalian cells.
  • 'Single-tube' system for reduced number of pipetting steps
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions or time-consuming alcohol precipitations
  • Reliable and absolutely reproducible procedure
  • High yields
  • Lysate can directly be used for genotyping PCRs
NOTE: 1 µl lysate is sufficient for subsequent genotyping PCR reactions.
Product Family Options
  • Descrizione
    Conf
  • DirectPCR®-Tail, optimised for lysis of mouse tail tissue, 50 ml
    50 mL
    1 * 50 mL
    1 * 50 mL - SFr. 287.00
    SFr. 287.00
    1 * 50 mL
       SFr. 287.00
  • DirectPCR®-Tail, optimised for lysis of mouse tail tissue, 100 ml
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 254.00
    SFr. 254.00
    1 * 100 mL
       SFr. 254.00
  • DirectPCR®-Yolk Sac, optimised for lysis of mouse embryonic material, 50 ml
    50 mL
    1 * 50 mL
    1 * 50 mL - SFr. 152.00
    SFr. 152.00
    1 * 50 mL
       SFr. 152.00
  • DirectPCR®-Yolk Sac, optimised for lysis of mouse embryonic material, 100 ml
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 254.00
    SFr. 254.00
    1 * 100 mL
       SFr. 254.00
  • DirectPCR®-Cell, optimised for lysis of mammalian cells, 50 ml
    50 mL
    1 * 50 mL
    1 * 50 mL - SFr. 152.00
    SFr. 152.00
    1 * 50 mL
       SFr. 152.00
  • DirectPCR®-Cell, optimised for lysis of mammalian cells, 100 ml
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 254.00
    SFr. 254.00
    1 * 100 mL
       SFr. 254.00
  • DirectPCR®-Ear, optimised for lysis of mouse ear tissue, 25 ml
    25 mL
    1 * 25 mL
    1 * 25 mL - SFr. 152.00
    SFr. 152.00
    1 * 25 mL
       SFr. 152.00
  • DirectPCR®-Ear, optimised for lysis of mouse ear tissue, 50 ml
    50 mL
    1 * 50 mL
    1 * 50 mL - SFr. 254.00
    SFr. 254.00
    1 * 50 mL
       SFr. 254.00
This product has been blocked by your organization. Please contact your purchasing department for more information