Gas detection tubes for hydrochloric acid
Nome del fornitore: Dräger
Certificati
Informazioni su questo articolo
To measure the 'spot concentrations' of hydrochloric acid in the air with an Accuro® pump - Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
- Hydrogen sulphide and sulphur dioxide do not interfere in the TLV range
- Impossible to measure hydrochloric acid in the presence of other mineral acids
- Chlorine and nitrogen dioxide are indicated but with different sensitivities
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Specifiche
- Catalog No:
- 118-0328
- 151-0152
- 151-0203
- Modello:
- Hydrochloric acid 0.2/a
- Hydrochloric acid 50/a
- Hydrochloric acid 1/a
- Modifica del colore:
- Blue to yellow
- Blue to pale yellow
- Blue to yellow
- Umidità:
- ≤ 15 mg/l H₂O
- ≤ 15 mg/l H₂O
- ≤ 15 mg/l H₂O
- Tempo di misurazione:
- 2 min/25 s
- 30 seconds/5 minutes
- 2 min
- N. di corse:
- 10/2
- 1/10
- 10
- Deviazione standard:
- ±10 - 15 %
- ±10 - 15 %
- ±10 - 15 %
- Gamma standard:
- 0,2 - 3/3 - 20 ppm
- 500 - 5000/50 - 500 ppm
- 1 - 10 ppm
- Range di temperatura:
- 5…40 °C
- 10…50 °C
- 5…40 °C
Specifiche
Informazioni correlate
Frequently Bought Together