Peqlab peqGOLD, Deoxynucleotide Triphosphate (Solution)
Nome del fornitore: VWR Peqlab
Certificati
Informazioni su questo articolo
Individually packaged deoxynucleotide triphosphates manufactured to the highest quality levels.
- In solution and ready to use, pH 7,5
- Synthetically manufactured
- Purity, measured by HPLC, >99%
- Highly stable, >90% stability after 6 weeks storage at room temperature
Certifications: Manufacturing and quality control under ISO 9001 and ISO 14001.
Informazioni correlate
Certificati
Cerca per numero di lotto20-101x dATP_d.pdf 20-101x dATP_e.pdf 20-102x dCTP_d.pdf 20-102x dCTP_e.pdf 20-104x dTTP_d.pdf
Scheda dati
Di più...
peqGOLD dNTPs_Lösungen.pdf
Letteratura sul prodotto
spec_PEQL20-1010 spec_PEQL20-1010 spec_PEQL20-1010 spec_PEQL20-1020 spec_PEQL20-1020
Specifiche del prodotto
Di più...
Frequently Bought Together