Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Anti-ACD Polyclonal Antibody Pair

Anti-ACD Polyclonal Antibody Pair

1 Set
Catalog # ABNOH00065057-PW2
Supplier:   Abnova
Anti-ACD Polyclonal Antibody Pair
Catalog # ABNOH00065057-PW2
Supplier:  Abnova
Anti-ACD Polyclonal Antibody Pair
Anti-ACD Polyclonal Antibody Pair

1 Set

Price
SFr. 992.00
Log in for availability
1 * 1 SET
Product Details & Documents

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ACD (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-ACD (50 µg)

Type:
Antigen: ACD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human

Specifications
  • Taglia:
    1 Set
  • Western blot:
    Yes
  • Cat. No.:
    ABNOH00065057-PW2
  • Reattività:
    Human
  • Sinomini antigene:
    TPP1|PIP1|TINT1|PTOP
  • Simbolo dell'antigene:
    ACD
  • Clonalità:
    Policlonale
  • Ospite:
    Multiple
  • ID gene:
    65057
  • ImmunoPrecipitazione:
    Yes
  • Temperatura di conservazione:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Tipo di anticorpi:
    Adrenocortical Dysplasia homolog
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..