Vacuum trap assembly, A584
Codice catalogo VARIA584
Nome del fornitore: VARIAN
Specifiche
- Descrizione:Vacuum trap assembly, includes 1 glass jar, 1 stopper, 2 elbow fittings and 2 tubes (I.Ø 6,35 mm, O.Ø 11,1 mm, length 457 mm)
- Environmentally Preferable:
- Conf:1
Specifiche
Informazioni su questo articolo
This vacuum trap eliminates the possibility of drawing fluids into the vacuum pump that may be detrimental to it.