Mobile phase inlet filter, Hastelloy® C
Mobile phase inlet filter with 1/8" tubing, 2 µm
Codice catalogo VARIAL9402
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
This mobile phase inlet filter operates up to 500 °C and is compatible with halogenated solvents.
Specifiche
- Porosità:2 µm
- Descrizione:Mobile phase inlet filter with 1/8" tubing