Galtek® check valves
Nome del fornitore: ENTEGRIS
Vai a
Informazioni su questo articolo
O-ring free design, ¹/₂ or ¹/₄" orifice.
- O-ring free design reduces contamination
- All wetted parts are molded PFA and PTFE for corrosion resistance
- No metal parts are used that could corrode or contribute to contamination
- Poppet design allows for a high flow through rate and maximises rinsability
6 varianti
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
- O-ring free design reduces contamination
- All wetted parts are molded PFA and PTFE for corrosion resistance
- No metal parts are used that could corrode or contribute to contamination
- Poppet design allows for a high flow through rate and maximises rinsability
Product Family Options
1/4" orifice
Informazioni sul prodotto
- DisponibilitàPrice
- ENTECK4-6F-6FInlet 3/8" Flaretek®, outlet 3/84" Flaretek®, length 89,7 mm"
- ENTECK4-4F-4FInlet 1/4" Flaretek®, outlet 1/4" Flaretek®, length 87,4 mm
- ENTECK4-4N-4QInlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm
- ENTECK4-4N-4NInlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm
1/2" orifice
Informazioni sul prodotto
- DisponibilitàPrice
- ENTECK8-8N-8NInlet 1/2" FNPT, outlet 1/2" FNPT, length 84,3 mm
- ENTECK8-8F-8FInlet 1/2" Flaretek®, outlet 1/2" Flaretek®, length 116,8 mm