Gowning bench
Nome del fornitore: Metro International
Vai a
Informazioni su questo articolo
Gowning bench in stainless steel 304.
- Heavy-duty, solid top bench provides comfortable gowning fixture
- Stainless steel foot plates provide floor mount or free-standing option
4 varianti
Product Details & Documents
Gowning bench in stainless steel 304.
- Heavy-duty, solid top bench provides comfortable gowning fixture
- Stainless steel foot plates provide floor mount or free-standing option
Documents
METOGB1636S_TDS_EN
Datasheet
Product Family Options
Informazioni sul prodotto
- PesoDisponibilitàPrice
- METOGB1636SGowning bench, L×W: 914×406 mm
- METOGB1648SGowning bench, L×W: 1219×406 mm
- METOGB1660SGowning bench, L×W: 1524×406 mm
- METOGB1672SGowning bench, L×W: 1825×406 mm