Dissolution vessels, QLA
Informazioni su questo articolo
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
- Made of high quality borosilicate glass or plastic
- Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
7 varianti
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
- Made of high quality borosilicate glass or plastic
- Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Product Family Options
Glass
Informazioni sul prodotto
- DisponibilitàPrice
- QLAAGLA900-01QLA clear glass dissolution vessel, serialised, 1000 ml
- QLAAGLA900-DKQLA clear glass dissolution vessel without ring, 1000 ml
- QLAAGLA900-EWQLA clear glass dissolution vessel, 1000 ml
- QLAAGLA900-HRQLA clear glass dissolution vessel, 1000 ml, Hanson compatible
- QLAAGLA900-VKQLA clear glass dissolution vessel, TruCenter, serialised, 1000 mlQLA clear glass dissolution vessel, TruCenter, serialised, 1000 mlLog in per disponibilità1 * 1 STSFr. 592.00
- QLAAGLA9CR-ADQLA amber glass dissolution vessel with centering ring, serialised, 1000 mlQLA amber glass dissolution vessel with centering ring, serialised, 1000 mlLog in per disponibilità1 * 1 STSFr. 654.00
- QLAAGLA9PK-01QLA clear glass dissolution Apex vessel, serialised, 1000 ml