Orders LinkContactUsLinkComponent
Transfection kit, BacMagic™
Transfection kit, BacMagic™
Novagen® BacMagic™ transfection kit, 5 reactions, 1 KIT
Codice catalogo 71546-3
Nome del fornitore:   Merck Millipore (Novagen)
Transfection kit, BacMagic™
Codice catalogo 71546-3
Nome del fornitore:  Merck Millipore (Novagen)
Numero CAS:  
Transfection kit, BacMagic™
Novagen® BacMagic™ transfection kit, 5 reactions, 1 KIT
Numero CAS:  
Price
1 360.00 SFr.
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
The Novagen® BacMagic™-1 transfection kit provides a positive selection for creating baculovirus recombinants in preparation for recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification.
The Novagen® BacMagic™-1 transfection kit contains the BacMagic™-1 DNA, Insect GeneJuice® transfection reagent and a positive control plasmid to monitor co-transfection efficiency. In addition, it contains Sf9 cells and BacVector® medium, for optimal transfection performance.
Specifiche
Specifiche
  • Conf:
    1 KIT
  • Descrizione:
    Novagen® BacMagic™ transfection kit, 5 reactions
  • Cat. No.:
    71546-3
  • Specifico per cella:
    Insect
  • Insetto:
    Yes