Orders LinkContactUsLinkComponent
Polycarbonate hydrometers
Polycarbonate hydrometers
Supplier:   Ludwig Schneider
Polycarbonate hydrometers
Supplier:  Ludwig Schneider
Certificates
Polycarbonate hydrometers
Polycarbonate hydrometers
2 Options Available Below
Product Details & Documents
These measuring devices are suitable for food without risk of glass breakage.
  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and high reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industry.
Reference temperature: 20 °C
Ordering information: Available with works or DAkkS calibration certificate.
Packaging: Individual square plastic sleeve with serial number.
Product Family Options
Product Information
  • Range di misura
    Lunghezza
    Availability
    Price
  • 1,200 - 1,420 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 96.00
    SFr. 96.00
    1 * 1 ST
       SFr. 96.00
  • 1,400 - 1,620 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 96.00
    SFr. 96.00
    1 * 1 ST
       SFr. 96.00