Orders LinkContactUsLinkComponent
Bandage mull
Bandage mull
Supplier:   HARTMANN PAUL
Bandage mull
Supplier:  HARTMANN PAUL
Certificates
Bandage mull
2 Options Available Below
Product Details & Documents
Mull for making individual bandages and wound dressings.
  • 20 threads
  • Laid 8-fold in zigzag layers to a width of 10 cm
  • Good purity and absorbency
Certifications: EN 14079
Product Family Options
Product Information
  • Descrizione
    Availability
    Price
  • Gauze, 20-ply, zigzag layers, 80 cm wide, 10 m long
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 44.90 SFr.
    44.90 SFr.
    1 * 8 ST
       44.90 SFr.
  • Gauze, 20-ply, zigzag layers, 80 cm wide, 40 m long
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176.00 SFr.
    176.00 SFr.
    1 * 1 ST
       176.00 SFr.